Anti-CD19 Mouse Monoclonal Antibody (Brilliant Violet® 785) [clone: HIB19]

Supplier: Biolegend
302239 302240
CA302239-BLEA 343.1 CAD
CA302239-BL CA10757-692
Anti-CD19 Mouse Monoclonal Antibody (Brilliant Violet® 785) [clone: HIB19]
Antibodies
Brilliant Violet 785™ anti-human CD19 [HIB19]; Isotype: Mouse IgG1, κ; Reactivity: Human, Chimpanzee, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: HIB19
Conjugation: Brilliant Violet® 785
Epitope:
Host: Mouse
Isotype:More Product Information
Order Now

SPECIFICATIONS

Antigen symbol CD19
Environmentally Preferable
Antigen synonyms B4
Antibody type Primary
Clonality Monoclonal
Conjugation Brilliant Violet® 785
Clone HIB19
Reactivity Rhesus monkey,Chimpanzee,Human
Host Mouse
Gene ID 930
Isotype IgG1 kappa
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
Purification The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 785™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 785™ and unconjugated antibody.
Storage temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR