UV5 Excellence UV/VIS Spectrophotometers, METTLER TOLEDO®
Supplier: Mettler-Toledo
30254725 30254725 30254728 30254728
10811-202EA 19655.2 CAD
10811-202 CA10811-200 CA10811-202 10811-200
UV5 Excellence UV/VIS Spectrophotometers, METTLER TOLEDO®
Spectrophotometers and Photometers UV/Visible Spectrophotometers
UV/VIS Excellence instruments effectively optimize spectroscopic workflows. FastTrack™ Technology allows for speedy measurements of the entire spectrum all within a compact footprint.
  • Unique system design with excellent optical performance
  • Xenon Flash Lamp combined with CCD array detector
  • No warm up time needed, the instrument is always ready for use
  • Open sample area for efficient cuvette handling
  • Robust design with no moving parts
  • Full spectrum scans and measurement results in as little as 3 seconds
UV/VIS Excellence instruments include One Click, an easy and intuitive way to run tasks right from the terminal.
The UV5Bio contains ready-to-use direct measurement applications and METTLER TOLEDO methods that cover a large selection of typical measuring modes and workflows for the Life Sciences including Nucleic Acid Analysis: 260/280nm ratios for nucleic acid purity. Protein Analysis: Biuret, Lowry, Bradford, bicinchoic acid and trinitrobenzene sulfonate protein quantification methods. Kinetics tests such as enzyme activity. Standard cuvette measurements available.
The UV5 is a simple and compact benchtop spectrophotometer with high level performance and flexibility in sample measurement. Extended pathlength capability as well as transparent solids and film analysis. A perfect solution for all general chemistry needs as well as extended color analysis for color scales such as APHA, Gardner, Saybolt, CIEL*a*b* and more.
Certifications: METTLER TOLEDO® provides ISO 9001 manufacturing, nationwide ISO 9001 field service, ISO 9001 quality certification, and ISO 14001 environmental certification. CE conformity mark provides you with the assurance that our products comply with the most recent EU directives. This product is CSA certified and UL listed.
Ordering information: Each instrument comes with a one-year manufacturer's warranty. Additional components, software, and accessories are available. Contact your Avantor sales representative for more information.