DuPont™ Proshield® 10 Frocks with Mandarin Collar, Elastic Wrists, and Zipper Closure
Supplier: DuPont
PB271SBU2X003000 PB271SBU3X003000 PB271SBU4X003000 PB271SBULG003000 PB271SBUMD003000 PB271SBUXL003000 PB271SWH2X003000 PB271SWH3X003000 PB271SWH4X003000 PB271SWHLG003000 PB271SWHMD003000 PB271SWHXL003000
10016-332CS 160.67 CAD
10016-332 10016-334 10016-336 10016-338 10016-340 10016-342 10016-344 10016-346 10016-348 10016-350 10016-352 10016-354
DuPont™ Proshield® 10 Frocks with Mandarin Collar, Elastic Wrists, and Zipper Closure
Disposable Garments
DuPont™ ProShield® 10, spunbond meltblown spunbond (SMS) garments, are made to the high quality standards you have come to expect from DuPont.
  • Mandarin collar
  • Elastic wrists
  • Front zipper closure
  • Length extends to knees
  • Serged seams
ProShield® 10 garments have been optimized for comfort, softness and breathability while enabling you to cut costs for disposable garments. DuPont™ ProShield® 10 disposable garments are designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications. Applications include general maintenance, janitorial/cleaning and other dirty work. Frocks provide body-length coverage with set sleeve design and front zipper closure for easy donning and doffing. Frocks feature a Mandarin collar, elastic wrists and serged seams.
Certifications: Thickness meets ASTM D1117.