DuPont™ Proshield® 10 Frocks with Mandarin Collar and Enclosed Elastic Wrists
Supplier: DuPont
ProShield®
PB267SWHMD003000 PB267SWH3X003000 PB267SWHSM003000 PB267SWHLG003000 PB267SWH4X003000 PB267SWHXL003000 PB267SWH2X003000
10016-330CS 204.52 CAD
10016-330 10016-320 10016-322 10016-324 10016-326 10016-328 10016-318
DuPont™ Proshield® 10 Frocks with Mandarin Collar and Enclosed Elastic Wrists
Disposable Garments
DuPont™ ProShield® 10, spunbond meltblown spunbond (SMS) garments, are made to the high quality standards you have come to expect from DuPont.
  • Mandarin collar
  • Enclosed elastic wrists
  • Set sleeve design
  • Knit cuffs
  • Length extends to knees
  • Front snap closure
  • Serged seams
ProShield® 10 garments have been optimized for comfort, softness and breathability while enabling you to cut costs for disposable garments. DuPont™ ProShield® 10 disposable garments are designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications. Applications include general maintenance, janitorial/cleaning and other dirty work. Frocks provide body-length coverage with set sleeve design and front snap closures (6+1 adjustable) for easy donning and doffing. Frocks feature a Mandarin collar, knit cuffs, enclosed elastic wrists and serged seams.
Certifications: Thickness meets ASTM D1117.