Roll-O-Matic® Self-Wringing Cleanroom Sponge Mops, Vileda Professional-FHP
Supplier: Vileda Professional - FHP
122910 118550 126104 118549 120848
94001-380EA 853.26 CAD
94001-380 94002-802 68400-492 89092-938 94002-800 89209-458
Roll-O-Matic® Self-Wringing Cleanroom Sponge Mops, Vileda Professional-FHP
Mops
The ideal self-wringing cleanroom mop
  • Straight handles allow user to properly use the pull-and-lift technique with both sides of the mop
  • Perfect for use on floors, walls, and ceilings
  • Ergonomic grips reduce worker fatigue
  • Entire aluminum and steel mop handles are completely autoclavable at the most common autoclave cycles
  • Caps on steel versions are stainless steel and completely sealed
  • Aluminum version features color coding tabs (blue, red, green, yellow) for easy equipment identification
The Vileda Professional® stainless steel and aluminum extendable Roll-O-Matic® mops are the best choice when seeking one mop to apply disinfectant, clean surfaces, or absorb spills in all production and controlled environments. These stainless steel Roll-O-Matic® mops are manufactured to resist repeated autoclave cycles and continued exposure to the common chemicals and disinfectants used in the controlled environment marketplace.