Stabilwax®-MS Columns (Fused Silica), Restek
Supplier: Restek
10673 10674
10849-668EA 1585.43 CAD
10849-668 10849-670
Stabilwax®-MS Columns (Fused Silica), Restek
Chromatography Columns
New Stabilwax®-MS columns ensure reproducible retention times from run to run, even with temperature cycling.
  • High-polarity, stable polyethylene glycol (PEG) stationary phase
  • Low bleed and rugged enough to withstand repeated temperature cycles without retention time shifting
  • Ideal for food, flavor, fragrance, and industrial chemical and solvent analysis
  • Temperature range: 40 to 250/260 °C
  • Equivalent to USP G14, G15, G16, G20, and G39 phases
When methods require trace analysis, this highly polar, low-bleed stationary phase produces excellent signal-to-noise levels. Ideal for food and flavor analysis (e.g., essential oils), fragrance and allergen analysis, as well as industrial solvent and chemical analysis.