Rxi®-PAH Columns (Fused Silica), Restek
Supplier: Restek
49316 49317 49318
10059-572EA 1935.18 CAD
10059-572 10059-574 10850-562
Rxi®-PAH Columns (Fused Silica), Restek
Chromatography Columns
The Rxi®-PAH GC columns were designed by Restek with a higher phenyl-content stationary phase that provides unique selectivity to separate important polycyclic aromatic hydrocarbons (PAH) for food safety that cannot be distinguished by mass spectrometry.
  • Ideal for EFSA PAH4 analysis—separates all priority compounds: benz[a]anthracene, chrysene, benzo[b]fluoranthene, and benzo[a]pyrene
  • Best resolution of chrysene from interfering PAHs, triphenylene, and cyclopenta[cd]pyrene
  • Complete separation of benzo [b], [k], [ j], and [a] fluoranthenes
  • 360°C thermal stability allows analysis of low-volatility dibenzo pyrenes
  • Mid-polarity proprietary phase
Even difficult priority compounds, such as the European Food Safety Authority (EFSA) PAH4, are easily separated and accurately quantified—results that cannot be achieved on typical GC columns. Arylene modification and surface bonding of the stationary phase increase thermal stability and ruggedness so relatively nonvolatile, higher molecular weight PAHs can be analyzed routinely without interference from column bleed. Excellent column efficiency means that the column can be trimmed for maintenance purposes many times without losing critical PAH separations, including those that are part of environmental methods, as well as food safety testing. The selectivity and efficiency of the Rxi®-PAH column make it ideal for EFSA PAH4 analysis; chrysene/triphenylene separation and resolution of all benzofluoranthenes are easily achieved.