Rtx®-BAC Plus 2 Columns (Fused Silica), Restek
Supplier: Restek
18006 18007
10058-506EA 1822.28 CAD
10058-506 10058-508
Rtx®-BAC Plus 2 Columns (Fused Silica), Restek
Chromatography Columns
These application-specific columns for blood alcohol analysis baseline separate all critical compounds—including ethanol, methanol, acetone, tert-butanol, acetaldehyde, isopropanol, and n-propanol—in less than 2 minutes.
  • Optimized column selectivities guarantee resolution of ethanol, internal standards, and frequently encountered interferences
  • Robust and reproducible column chemistry ensures longer column lifetime and consistent results
  • Stable to 260 °C
Every Rtx®-BAC Plus 1 and Rtx®-BAC Plus 2 column is qualified with a test mix containing theseMore Product Information
Order Now
SPECIFICATIONS
Description 10058-506 10058-508
Environmentally Preferable
Temperature limit -20 to 240/260 °C -20 to 240/260 °C