Stabilwax® Secondary Columns for GCxGC (fused silica), Restek
Supplier: Restek
15112 15118 15125
10850-496EA 215.65 CAD
10850-496 10850-498 10850-500
Stabilwax® Secondary Columns for GCxGC (fused silica), Restek
Chromatography Columns
Restek’s polar-deactivated surface tightly binds the Carbowax® polymer and increases thermal stability, relative to competitive columns.
  • Convenient 2 m length is ideal for use as a secondary column in GCxGC analyses
  • Most stable polyethylene glycol (PEG) phase available
  • Rugged enough to withstand repeated water injections
  • Temperature range: 40 to 260 °C
  • Polar phase; Crossbond® polyethylene glycol
Because of the increased stability produced by the bonding process, Stabilwax® columns exhibit long column lifetime, even when programming repeatedly up to 260More Product Information
Order Now
SPECIFICATIONS
Description 10850-496 10850-498 10850-500
Environmentally Preferable
Temperature limit 40 to 250/260 °C 40 to 250/260 °C 40 to 250/260 °C