Rxi®-1ms Secondary Columns for GCxGC (fused silica), Restek
Supplier: Restek
15114 15120 15127
10850-672EA 215.65 CAD
10850-672 10850-674 10850-676
Rxi®-1ms Secondary Columns for GCxGC (fused silica), Restek
Chromatography Columns
Rxi® fused silica capillary GC columns are designed specifically for exceptional inertness, ultra-low bleed, and reproducible retention and selectivity.
  • Convenient 2 m length is ideal for use as a secondary column in GCxGC analyses
  • General-purpose columns
  • Temperature range: −60 to 330/350 °C (bleed tested temperature/maximum operating temperature)
  • Ultra-low bleed
  • Exceptional inertness
  • Reproducible performance, column after column
  • Nonpolar phase; Crossbond® dimethyl polysiloxane
In addition to outstanding performance, this combination of columnMore Product Information