Rtx®-Volatile Amine Columns (Fused Silica), Restek
Supplier: Restek
18076 18077 18078
10058-510EA 1209.19 CAD
10058-510 10058-512 10058-514
Rtx®-Volatile Amine Columns (Fused Silica), Restek
Chromatography Columns
The Rtx®-Volatile Amine column was designed specifically for analyzing volatile amines in difficult matrices, such as water.
  • Unique selectivity for baseline resolution of all volatile amines
  • Excellent inertness assures accuracy and sensitivity for volatile amines, including free ammonia
  • Highly robust phase withstands repeated water injections, resulting in longer column lifetime
  • High temperature stability (290 °C) ensures elution of amines up to C16 and allows contaminants to be removed by 'baking out' the column
The unique base deactivation creates an exceptionally inert surface for these sensitive compounds, resulting in highly symmetrical peaks, which allow low detection limits.
It is recommended to use base-deactivated fused silica guard columns and base-deactivated liners with Rtx®-Volatile Amine columns.
The stable bonded phase yields a column that is not only retentive and highly selective for these compounds but is also very robust and able to withstand repeated water injections. Comparisons made by customers performing routine volatile amine applications have shown the Rtx®-Volatile Amine column outperforms other amine-specific columns, especially for peak shape and lifetime. Each Rtx®-Volatile Amine column is held to stringent quality specifications and tested with a specially designed test mix that includes basic compounds to ensure exceptional inertness, reliability, and reproducibility. These qualities assure consistent performance and make the Rtx®-Volatile Amine column the best choice for volatile amines analysis.