2abc Refinery Gas Column Set (packed), Restek
Supplier: Restek
88000-810 88000-875 88004-875
10848-676EA 2404.76 CAD
10848-676 10848-682 10848-692
2abc Refinery Gas Column Set (packed), Restek
Chromatography Columns
This 3-column set is finely tuned to resolve light hydrocarbons.
  • Special columns for unsaturated light hydrocarbons
  • Faster separations of C1 to C4 hydrocarbons
  • To be used with 2abc refinery gas column set to backflush and prevent C6+ hydrocarbons from entering column set
When used in the proper valving system, it will elute C5+ hydrocarbons ahead of C1 through C4 hydrocarbons.