Rxi®-1HT Columns (Fused Silica), Restek
Supplier: Restek
13951 13950 13952 13953 13954 13955
89205-678EA 1407.13 CAD
89205-678 10058-482 10850-478 10850-480 10850-482 10850-484
Rxi®-1HT Columns (Fused Silica), Restek
Chromatography Columns
Rxi® fused silica capillary GC columns are designed specifically for exceptional inertness, ultra-low bleed, and reproducible retention and selectivity.
  • 40% longer lifetime from specially designed fused silica tubing
  • Columns processed for high-temperature applications, such as high molecular weight hydrocarbons
  • Temperature range: −60 to 400 °C
  • Nonpolar phase; dimethyl polysiloxane
In addition to outstanding performance, this combination of column characteristics assures Rxi® columns will have long lifetimes. Restek has also developed a stable, reproducible manufacturing process, so that Rxi® columns will meet your needs every time you install a new column.