Rxi®-17Sil MS Columns (Fused Silica), Restek
Supplier: Restek
14123 14102 14120 14121 14124 14126 43820 43821 14111
89205-682EA 1869.98 CAD
89205-682 10058-484 10058-486 10058-488 10058-490 10058-492 10059-560 10059-562 10849-704
Rxi®-17Sil MS Columns (Fused Silica), Restek
Chromatography Columns
Rxi® fused silica capillary GC columns are designed specifically for exceptional inertness, ultra-low bleed, and reproducible retention and selectivity.
  • Excellent inertness and selectivity for active environmental compounds, such as PAHs
  • Low bleed for use with sensitive detectors, such as MS
  • 340/360 °C upper temperature limits
  • Equivalent to USP phase G3
  • Mid-polarity Crossbond® phase
In addition to outstanding performance, this combination of column characteristics assures Rxi® columns will have long lifetimes. Restek has also developed a stable, reproducible manufacturing process, so that Rxi® columns will meet your needs every time you install a new column.