Rxi®-624Sil MS Columns (Fused Silica), Restek
Supplier: Restek
13868 13865 13869 13870 13871 13872 13873 13874 13875
89205-680EA 1627.3 CAD
89205-680 89421-664 10058-470 10058-472 10058-474 10058-476 10058-478 10058-480 10849-576
Rxi®-624Sil MS Columns (Fused Silica), Restek
Chromatography Columns
Rxi® fused silica capillary GC columns are designed specifically for exceptional inertness, ultra-low bleed, and reproducible retention and selectivity.
  • Low-bleed, high-thermal stability column—maximum temperatures up to 300 to 320 °C
  • Inert—excellent peak shape for a wide range of compounds
  • Selective—G43 phase highly selective for volatile organics and residual solvents, great choice for USP 467
  • Manufactured for column-to-column reproducibility—well-suited for validated methods
  • Mid-polarity Crossbond® phase
In addition to outstanding performance, thisMore Product Information