MXT®-Msieve 5A Columns (Siltek®-treated stainless steel PLOT), Restek
Supplier: Restek
79723 79723-273
10850-882EA 2026.75 CAD
10850-882 10851-054 10851-118 10851-204
MXT®-Msieve 5A Columns (Siltek®-treated stainless steel PLOT), Restek
Chromatography Columns
Restek’s molecular sieve 5A PLOT columns are designed for efficient separation of argon/oxygen and other permanent gases, including carbon monoxide.
  • Advantages of metal MXT® PLOT columns include:
  • Can be made in small coil diameters—perfect for tight spaces
  • Rugged material withstands rough handling and shock
  • Designed for robust performance in GC processes and field instruments
  • Available in 3.5"-coil diameter or 7"-diameter, 11-pin cage
Special coating and deactivation procedures ensure chromatographic efficiency and the integrity of the porous layer coating. Molecular sieves have very high retention, allowing separations of permanent gases at temperatures above ambient. Restek's deactivation technology also allows carbon monoxide to elute as a sharp peak. Additionally, Restek's unique immobilization process guarantees that the uniform particles remain adhered to the tubing—even after continuous valve cycling.