PLOT Column Particle Trap, Restek
Supplier: Restek
19754 19753
10059-034EA 379.53 CAD
10059-034 10058-554
PLOT Column Particle Trap, Restek
Chromatography Columns
PLOT column particle traps are effective for eliminating detector spikes and scratches in valve rotors.
  • Includes two Press-Tight® connectors and a 2.5 m column
  • Protects detector and valves; connects between column and detector or valve
  • Eliminates detector spikes and scratches in valve rotors
The technology used to adhere particles in PLOT columns is excellent; however, it is still possibile for particles to dislodge when extreme pressure shocks and gas flow changes occur. This sometimes happens when valve switching or backflushing is used. In thoseMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable