D3606 Application Column Set (packed, 2 column set), Restek
Supplier: Restek
83606-800 83606-800901 83606-810 83606-810901 83606-840 83606-850
10848-656EA 1849.43 CAD
10848-656 10848-658 10848-660 10848-662 10848-668 10848-670
D3606 Application Column Set (packed, 2 column set), Restek
Chromatography Columns
D3606 application column sets are thoroughly tested for method applicability and the accurate quantification of benzene and toluene.
  • Complete resolution of benzene from ethanol—no compromising coelutions
  • Accurate quantification of benzene and toluene
  • Column sets are individually tested for method applicability
  • Listed in the appendix of ASTM Method D3606 as an acceptable alternative to TCEP columns—get better separation of benzene and ethanol while still following ASTM method requirements
The column set is designed to accommodate both valve injectionMore Product Information