Rxi® Guard/Retention Gap Columns (fused silica), Restek
Supplier: Restek
10054-600 10073-600 10039 10054 10059 10064 10073 10029 10059-600 10064-600 10029-600 10039-600
10058-148PK 874.9 CAD
10058-148 10058-150 10059-002 10059-004 10059-006 10059-008 10059-010 10803-186 10852-102 10852-104 10852-106 10852-108
Rxi® Guard/Retention Gap Columns (fused silica), Restek
Chromatography Columns
Rxi® guard/retention gap columns are specially designed to be exceptionally inert.
  • Extend column lifetime
  • Excellent inertness-obtain lower detection limits for active compounds
  • Sharper chromatographic peaks by utilizing retention gap technology
  • Maximum temperature: 360 °C
These columns are fabricated from durable fused silica.