CarboBlack Columns (packed), Restek
Supplier: Restek
80122-800 80205-810 80121-800 80205-840
RK80122-800EA 390.65 CAD
RK80122-800 RK80205-810 RK80121-800 10803-498 10847-962 10847-964 10848-168
CarboBlack Columns (packed), Restek
Chromatography Columns
Graphitized carbon black offers unique selectivity and very little adsorption for alcohol analyses.
  • Siltek®-treated stainless steel
  • CarboBlack C model
Two types of CarboBlack supports are available, CarboBlack B and CarboBlack C. CarboBlack B supports, with their higher surface area, can hold up to a 10% loading of a nonsilicone liquid phase. CarboBlack C supports can hold up to a 1% loading of a nonsilicone liquid phase. Many Carbowax® 20M-loaded CarboBlack packings are available. CarboBlack packings are treated with KOH or picric acid for basic or acidic compounds, and special alcoholic beverage loadings are available. CarboBlack supports provide resolution and retention similar to Carbopack™ and Carbograph supports.