Molecular Sieve Columns (packed), Restek
Supplier: Restek
80428-810 80473-810 80474-810 80475-810 80473-800 88015-810 80428-800 80428-840 80429-800 80429-810 80429-820 80429-840 80430-800 80430-810 80439-800 80439-810 80439-820 80440-800 80440-810 80474-800 80474-840 80475-800 80475-820 80476-800 80476-810 80476-820 80480-800 88014-800 88014-810 88015-800
10058-316EA 456.74 CAD
10058-316 10058-320 10058-322 10058-324 10803-500 10803-504 10848-276 10848-282 10848-288 10848-292 10848-298 10848-300 10848-308 10848-312 10848-366 10848-368 10848-372 10848-376 10848-378 10848-530 10848-538 10848-546 10848-550 10848-556 10848-560 10848-564 10848-590 10848-718 10848-722 10848-736
Molecular Sieve Columns (packed), Restek
Chromatography Columns
Molecular sieve packed columns easily separate permanent gases at above-ambient temperatures.
  • Siltek®-treated stainless steel
  • Preactivated and ready to use
  • Excellent batch-to-batch reproducibility
Restek has developed a process for preparing molecular sieve packings, which results in excellent batch-to-batch reproducibility. In addition, these molecular sieves are preactivated and ready to use. Each column comes with metal end-fittings to prevent water or carbon dioxide from adsorbing into the packing during shipment.