Chromosorb® Diatomaceous Earth Columns (bonded stationary phase packed columns), Restek
Supplier: Restek
80405-810 80409-800 80409-810 80448-800 80449-810 80441-800 80442-800 80442-810 80442-840 80443-800 80443-810 80447-800
RK80405-810EA 412.69 CAD
RK80405-810 RK80409-800 RK80409-810 RK80416-800 RK80401-810 RK80415-840 RK80416-810 RK80416-840 10847-934 10847-938 10847-942 10848-114 10848-116 10848-388 10848-396 10848-398 10848-402 10848-404 10848-406 10848-424 10848-428 10848-576 10848-580 10848-582
Chromosorb® Diatomaceous Earth Columns (bonded stationary phase packed columns), Restek
Chromatography Columns
Bonded phases are more resistant to oxidation than non-bonded phases because of the stronger intermolecular forces produced by cross-linking.
  • Low bleed levels
  • Longer column lifetime
  • Short conditioning times
  • Siltek®-treated stainless steel
  • Modified version of Chromosorb W; highest inertness, most consistent performance
  • Application specific column
Bonded phases are used in capillary columns because they provided a dramatic increase in column quality.
Solvent rinsing during the manufacturing process removes low molecular weight polymer fragments that contribute to long conditioning cycle times. Restek's bonded packings usually stabilize in 30 minutes or less. Because the bleed is lower with bonded phases, detectors can be operated at higher sensitivities and the need for data system bleed compensation is greatly reduced.
Now packed column chromatographers can expect shorter conditioning times, lower bleed, and longer column lifetime by using Restek bonded phase packed columns. Because the material is thoroughly cross-linked, the phase will not sag, as often happens with non-bonded phases.