Rxi®-17 Columns (Fused Silica), Restek
Supplier: Restek
13501 13502 13520 13523 13524 13525 13538 13539 13540 13552 13555 13570 13553 13554
10058-388EA 1237.12 CAD
10058-388 10058-390 10058-392 10058-394 10058-396 10058-398 10058-400 10058-402 10058-404 10058-406 10058-408 10058-412 10850-804 10850-806
Rxi®-17 Columns (Fused Silica), Restek
Chromatography Columns
Rxi® fused silica capillary GC columns are designed specifically for exceptional inertness, ultra-low bleed, and reproducible retention and selectivity.
  • General-purpose columns for pesticides, herbicides, rosin acids, phthalate esters, triglycerides, sterols
  • Temperature range: 40 to 320 °C
  • Mid-polarity phase; Crossbond® diphenyl dimethyl polysiloxane
This combination of column characteristics assures Rxi® columns will have long lifetimes. A stable, reproducible manufacturing process ensures that Rxi® columns will meet your needs every time you install a new column.