Rtx®-PCB Columns (Fused Silica), Restek
Supplier: Restek
13223 13226 13239 41302 41303 41304
10850-446EA 1293.81 CAD
10850-446 10850-448 10850-450 10850-536 10850-538 10850-540
Rtx®-PCB Columns (Fused Silica), Restek
Chromatography Columns
Rtx®-PCB columns are uniquely designed for PCBs analysis by GC-ECD or GC-MS while providing good results for other semi-volatiles.
  • Unique polymer for PCBs analysis by GC-ECD or GC-MS
  • Good results for other semi-volatiles
  • Low polarity; inert to active compounds
  • Stable to 340 °C
  • Proprietary Crossbond® phase
These columns are constructed from fused silica and are inert to active compounds.