Stx®-CLPesticides Columns (fused silica with Siltek® deactivation), Restek
Supplier: Restek
11546 11540 11541 11543 11544-600
10849-488EA 1429.02 CAD
10849-488 10850-396 10850-398 10850-400 10852-000
Stx®-CLPesticides Columns (fused silica with Siltek® deactivation), Restek
Chromatography Columns
Siltek® passivation technology enables these columns to offer unsurpassed inertness and the highest responses for active pesticides.
  • Application-specific columns for organochlorine pesticides and herbicides
  • Baseline separations in less than 10 minutes
  • Siltek® surface deactivation enhances responses for endrin, DDT, methoxychlor
  • Stable to 330 °C
  • Proprietary Crossbond® phases
Many laboratories analyzing organochlorine pesticides struggle with breakdown and adsorption of endrin, DDT, and methoxychlor caused by active sites throughout the analytical system, problems which are addressed by these columns.