MXT®-502.2 Columns (Siltek®-treated stainless steel), Restek
Supplier: Restek
70910 70919 70908
RK70910EA 3337.59 CAD
RK70910 10849-034 10851-058
MXT®-502.2 Columns (Siltek®-treated stainless steel), Restek
Chromatography Columns
An MXT®-502.2 column will enable you to quantify all compounds listed in U.S.
  • Application-specific columns with unique selectivity for volatile organic pollutants
  • Excellent separation of trihalomethanes; ideal polarity for light hydrocarbons and aromatics
  • Temperature range: −20 to 320 °C
  • 4.5" standard coil diameter
  • Proprietary Crossbond® diphenyl/dimethyl polysiloxane phase
The diphenyl/dimethyl polysiloxane based MXT®-502.2 stationary phase provides low bleed and thermal stability to 320 °C. A 105 m column can separate the light gases specified in EPA methods without subambient cooling.
EPA methods 502.2 or 524.2, whether you use a mass spectrometer or a PID in tandem with an ELCD.