MXT®-WAX Columns (Siltek®-treated stainless steel), Restek
Supplier: Restek
70670 70655 70651 70658 70666 70667 70620 70622 70623 70624 70625 70637 70638 70639 70640 70642 70652 70654 70657 70669 70672
RK70670EA 1656.03 CAD
RK70670 RK70655 10848-944 10848-946 10850-932 10851-066 10851-068 10851-072 10851-074 10851-250 10851-444 10851-570 10851-574 10851-576 10851-578 10851-580 10851-584 10851-586 10851-588 10851-590 10851-592 10851-596 10851-600 10851-606 10851-612 10851-614
MXT®-WAX Columns (Siltek®-treated stainless steel), Restek
Chromatography Columns
MXT®-WAX columns are good for general-purpose applications involving FAMEs, flavor compounds, essential oils, amines, solvents, xylene isomers, and U.S.
  • General-purpose columns for FAMEs, flavor compounds, essential oils, amines, solvents, xylene isomers, and U.S. EPA method 603 (acrolein/acrylonitrile)
  • Equivalent to USP G14, G15, G16, G20, and G39 phases
  • 4.5" standard coil diameter
  • Polar phase; Crossbond® Carbowax® polyethylene glycol—provides oxidation resistance
These columns are constructed of Siltek®-treated stainless steel.
EPA method 603.