MXT®-1701 Columns (Siltek®-treated stainless steel), Restek
Supplier: Restek
72040 72023 72070 72052 72053 72055 72082 72085 72088
RK72040EA 1674.28 CAD
RK72040 10849-046 10851-060 10851-522 10851-524 10851-528 10851-534 10851-538 10851-542
MXT®-1701 Columns (Siltek®-treated stainless steel), Restek
Chromatography Columns
MXT®-1701 columns are good for general-purpose applications involving alcohols, oxygenates, PCB congeners, and pesticides.
  • General-purpose columns for alcohols, oxygenates, PCB congeners (e.g., Aroclor mixes), and pesticides
  • Equivalent to USP G46 phase
  • 4.5" standard coil diameter
  • Mid-polarity Crossbond® phase
These columns are constructed of Siltek®-treated stainless steel.