Rtx®-20 Columns (Fused Silica), Restek
Supplier: Restek
10324 10339 10353 10355 10323 10354 10338
RK10324EA 1259.06 CAD
RK10324 RK10339 RK10353 RK10355 RK10323 RK10354 RK10338
Rtx®-20 Columns (Fused Silica), Restek
Chromatography Columns
Rtx®-20 polymer is synthesized to exacting standards so that all residual catalysts and low molecular weight fragments are removed from the polymer, providing a tight mono-modal distribution and extremely low bleed.
  • General-purpose columns for volatile compounds, flavor compounds, alcoholic beverages
  • Temperature range: −20 to 320 °C
  • Equivalent to USP G28, G32 phases
  • Low- to mid-polarity phase; Crossbond® diphenyl dimethyl polysiloxane
Rtx®-20 columns are made of fused silica and feature extremely low bleed levels.