Deactivated Fused Silica Tubing (Nonpolar), Restek
Supplier: Restek
10010 10020 10009 10011 10021 10031 10030
RK10010EA 646.91 CAD
RK10010 RK10020 RK10009 RK10011 RK10021 RK10031 RK10030
Deactivated Fused Silica Tubing (Nonpolar), Restek
Chromatography Columns
This fused silica tubing is commonly used as guard columns, transfer lines, or for long retention gaps.
  • Inertness tested with the Grob test mix
  • 5-meter banded sections mounted in a Restek cage
  • Maximum temperature: 325 °C
  • Note: 30 and 60 m lengths are banded in 5 m sections
These lengths of tubing are nonpolar and provided in 5 m banded sections.