Rt®-βDEXcst Columns (Fused Silica), Restek
Supplier: Restek
13102 13103
10848-750EA 1781.77 CAD
10848-750 10849-454
Rt®-βDEXcst Columns (Fused Silica), Restek
Chromatography Columns
By adding β or γ cyclodextrin to Restek's bonded Rtx®-1701 stationary phase, we greatly enhance overall utility and column lifetime for Restek's chiral columns, compared to columns that have pure cyclodextrin stationary phases.
  • Proprietary cyclodextrin material added into 14% cyanopropylphenyl/86% dimethyl polysiloxane
  • Improved column lifetime
  • Enhanced column utility
Separations of more than one hundred chiral compounds have been achieved using Restek's unique DEX columns, and Restek's columns continue to demonstrate stability after hundreds ofMore Product Information