Rt®-βDEXsa Columns (fused silica), Restek
Supplier: Restek
13108 13109
10849-508EA 1781.77 CAD
10849-508 10849-510
Rt®-βDEXsa Columns (fused silica), Restek
Chromatography Columns
By adding β or γ cyclodextrin to Restek's bonded Rtx®-1701 stationary phase, we greatly enhance overall utility and column lifetime for Restek's chiral columns, compared to columns that have pure cyclodextrin stationary phases.
  • 2,3-di-acetoxy-6-O-tert-butyl dimethylsilyl beta cyclodextrin added into 14% cyanopropylphenyl/86% dimethyl polysiloxane
  • Improved column lifetime
  • Enhanced column utility
Separations of more than one hundred chiral compounds have been achieved using Restek's unique DEX columns, and Restek's columns continue to demonstrateMore Product Information
Order Now
SPECIFICATIONS
Description 10849-508 10849-510
Environmentally Preferable
Temperature limit 40 to 230 °C 40 to 230 °C