Rtx®-Volatiles Columns (Fused Silica), Restek
Supplier: Restek
10905 10902 10903 10901 10900 10904 10906
RK10902EA 1485.69 CAD
RK10902 RK10901 RK10900 RK10906 RK10905 RK10904 RK10903
Rtx®-Volatiles Columns (Fused Silica), Restek
Chromatography Columns
Rtx®-Volatiles columns were the first columns designed specifically for analyses of the 34 volatile organic pollutants listed in U.S.
  • Application-specific columns for volatile organic pollutants
  • Stable to 280°C
  • Proprietary Crossbond® diphenyl/dimethyl polysiloxane phase
With these columns, you can quantify all compounds listed in these methods, whether you use a mass spectrometer or a PID in tandem with an ELCD. The diphenyl/dimethyl polysiloxane based Rtx®-Volatiles stationary phase provides low bleed and thermal stability to 280°C. Narrow bore columns can interface directly in GC/MS systems.
EPA methods 601, 602, and 624.