HayeSep® Packing Materials, Restek
Supplier: Avantor
25032
25033
25036
25037
25038
25039
25045
25046
25047
25048
25050
25053
25054
25055
25056
25563
25568
25569
25570
25571
25572
25574
25575
10847-852EA
379.44
CAD
10847-852
10847-858
10847-860
10847-866
10847-868
10847-870
10847-872
10847-878
10847-880
10847-882
10847-884
10847-886
10847-888
10847-890
10847-894
10847-896
10847-898
10847-900
10848-014
10848-024
10848-026
10848-028
10848-030
10848-032
10848-034
10848-036
10848-038
HayeSep® Packing Materials, Restek
Chromatography Sorbents
GC Sorbents
HayeSep® porous polymers are macroporous, spherical, ultrapure resins for performing unique separations in gas chromatography.
- Thoroughly cleaned and preconditioned
- No shrinkage
- Low bleed
Columns packed with HayeSep® polymers produce lower bleed and more reproducible characteristics than those packed with any competing polymeric product, and they require minimal conditioning.