MXT®-1 Columns (Siltek®-treated stainless steel), Restek
Supplier: Restek
70122 70109 70137 70124 70142 70121 70135 70139 70129 70105 70106 70107 70116 70120 70123 70125 70126 70128 70138 70140 70143 71811 71812 71815 70102 70105-600
RK70122EA 979.99 CAD
RK70122 RK70101 RK70109 RK70137 RK70124 RK70142 RK70121 10848-912 10848-918 10848-958 10848-976 10850-894 10850-942 10851-024 10851-080 10851-082 10851-334 10851-336 10851-368 10851-656 10851-660 10851-662 10851-714 10851-716 10851-720 10851-722 10851-724 10851-728 10851-730 10851-732 10851-736 10851-738 10851-740 10851-742 10851-746 10851-854 10851-856 10851-860 10851-880 10851-888 10852-126 10852-146 75849-436 75849-488 75849-586 75849-746 75850-198 75850-294 75850-496 75850-830 75850-832 75852-300
MXT®-1 Columns (Siltek®-treated stainless steel), Restek
Chromatography Columns
MXT®-1 columns exhibit long lifetime and very low bleed at high operating temperatures.
  • General-purpose columns for solvent impurities, PCB congeners (e.g., Aroclor mixes), gases, natural gas odorants, sulfur compounds, essential oils, hydrocarbons, semi-volatiles, pesticides, and oxygenates
  • Equivalent to USP G1, G2, G38 phases
  • 4.5" standard coil diameter
  • Nonpolar phase; Crossbond® dimethyl polysiloxane
A proprietary synthesis process eliminates residual catalysts that could cause degradation and increase bleed.