CarboBlack Packing Materials, Restek
Supplier: RESTEK CORP
25500 25501 25502 25503 25507 25508 25510 25511
10847-910EA 56.05 CAD
10847-910 10847-912 10847-986 10847-988 10847-990 10847-992 10847-994 10847-996 10847-998 10848-000
CarboBlack Packing Materials, Restek
Chromatography Sorbents GC Sorbents
Graphitized carbon black offers unique selectivity and very little adsorption for alcohol analyses.
  • CarboBlack B supports up to 10% loading of a nonsilicone liquid phase
  • CarboBlack C supports up to 1% loading of a nonsilicone liquid phase
Two types of CarboBlack supports are available, CarboBlack B and CarboBlack C. CarboBlack B support, with its higher surface area, can hold up to a 10% loading of a nonsilicone liquid phase. CarboBlack C support can hold up to a 1% loading of a nonsilicone liquid phase. Many Carbowax® 20M-loaded CarboBlack packings are available. CarboBlack packings are treated with KOH or picric acid for basic or acidic compounds, and special alcoholic beverage loadings are available. CarboBlack supports provide resolution and retention similar to Carbopack™ and Carbograph supports.
Ordering information: Minimum order of 10 grams. Price is per gram.