Rt®-Msieve 5A Columns (fused silica PLOT), Restek
Supplier: Restek
19720 19721 19722 19723 19773
10058-276EA 1686.41 CAD
10058-276 10058-280 10058-530 10058-532 10058-534 10058-536 10848-938 10848-940 75849-762 75849-848 75849-960
Rt®-Msieve 5A Columns (fused silica PLOT), Restek
Chromatography Columns
Restek’s molecular sieve 5A PLOT columns are designed for efficient separation of argon/oxygen and other permanent gases, including carbon monoxide.
  • Improve accuracy with sharp, symmetrical peaks for argon, oxygen, and carbon monoxide
  • Easily separate permanent gases at temperatures above ambient
  • Restek PLOT technology reduces particle release, improving flow reproducibility and reducing downtime for maintenance
  • Stable to 300 °C
Special coating and deactivation procedures ensure chromatographic efficiency and the integrity of the porous layer coating.More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Temperature limit 300 °C