Rtx®-BAC2 Columns (Fused Silica), Restek
Supplier: Restek
18000 18002
RK18000EA 1346.85 CAD
RK18000 10803-202 10849-280 10850-082
Rtx®-BAC2 Columns (Fused Silica), Restek
Chromatography Columns
These columns separate to baseline all blood alcohol compounds in blood, breath, or urine, in less than 3 minutes, under isothermal conditions.
  • Application-specific columns for blood alcohol analysis
  • Also excellent for abused inhalant anesthetics, γ-hydroxybutyrate (GHB)/γ-butyrolactone (GBL), glycols, and common industrial solvents
  • Rtx®-BAC2 confirmation column provides four elution order changes under the same conditions
  • Stable to 260 °C
  • Proprietary Crossbond® phase
Isothermal analysis increases productivity by eliminating the need for oven cycling.More Product Information
Order Now
SPECIFICATIONS
Description RK18000 10803-202
Environmentally Preferable
Temperature limit -20 to 240/260 °C -20 to 240/260 °C –20...240/260 °C