Rtx®-BAC1 Columns (Fused Silica), Restek
Supplier: Restek
18001 18003 18001-126
RK18001EA 1671.01 CAD
RK18001 10849-128 10849-156 10849-420 10849-908
Rtx®-BAC1 Columns (Fused Silica), Restek
Chromatography Columns
These columns separate to baseline all blood alcohol compounds in blood, breath, or urine, in less than 3 minutes, under isothermal conditions.
  • Application-specific columns for blood alcohol analysis
  • Also excellent for abused inhalant anesthetics, γ-hydroxybutyrate (GHB)/γ-butyrolactone (GBL), glycols, and common industrial solvents
  • Rtx®-BAC2 confirmation column provides four elution order changes under the same conditions
  • Stable to 260 °C
  • Proprietary Crossbond® phase
Isothermal analysis increases productivity by eliminating the need for oven cycling. Confirmation is easily achieved with this tandem set because there are four elution order changes between the two columns.