Base-Deactivated Guard/Retention Gap Columns (fused silica), Restek
Supplier: Restek
10001 10000 10002 10000-600 10002-600 10001-600
RK10001EA 169.96 CAD
RK10001 RK10000 RK10002 10849-944 10849-946 10850-160
Base-Deactivated Guard/Retention Gap Columns (fused silica), Restek
Chromatography Columns
Base-deactivated guard/retention gap columns are tested with a basic amine test mix.
  • Excellent inertness for basic compounds
  • Recommended for use with Rtx®-5 Amine, Rtx®-35 Amine, Rtx®-Volatile Amine, and Stabilwax®-DB capillary columns
  • Batch test chromatogram included
  • Maximum temperature: 315 °C
Chemists using guard columns in the analyses of basic compounds frequently observe peak tailing and low recovery. This happens because conventionally deactivated tubing surfaces can be adsorptive to basic compounds. Restek offers base-deactivated guard columns, as well as base-deactivated inlet liners, for completely inert sample pathways.