Pressure Refill Reaction Vials, Chemglass
Supplier: Chemglass
CG-4912-01 CG-4912-02 CG-4912-05 CG-4912-06
89171-852PK 362.8 CAD
89171-852 89171-854 89171-856 89171-858
Pressure Refill Reaction Vials, Chemglass
Vials
Made from 33 expansion borosilicate glass, ideal for use in heated reactions.
  • High resistance to thermal shock
  • Maximum chemical resistance
Certifications: Conforms to ASTM E-438 Type 1-Class A, USP Type 1 and ISO 3585.
Delivery information: Supplied with a special open top cap which prevents the pressure in the vial from exceeding 150 psig.
Packaging: Packaged in a SBS coated white tray complete with SBS partition. Boxes are marked with alpha-numeric coordinates for fast and easy vial identification.