Nalgene® Wash Bottle, FEP, Right-to-Understand, Thermo Scientific
Supplier: Thermo Fisher Scientific
2429-0502 2429-0503 2429-0504 2429-0505 2429-0506 2429-0501
10024-301CS 2489.88 CAD
10024-301 10024-303 10024-305 10024-307 10024-309 10024-805
Nalgene® Wash Bottle, FEP, Right-to-Understand, Thermo Scientific
Bottles Wash Bottles
New globally harmonized chemical labeling rules have been enacted to protect workers who come into contact with hazardous chemicals.
  • Bottles are made from high-purity FEP for low extractables and long life in the lab, and are suitable for dispensing high-purity solvents
  • Permanently laser etched with 100% Globally Harmonized System (GHS) chemical safety information
  • No other system markings are used on the bottles, which eliminates confusion
  • Safety information is laser etched directly into the wall of the bottle so information will remain permanent and
More Product Information