Super Nuova+™ Hot Plate Stirrer, Thermo Scientific
Supplier: Thermo Fisher Scientific
SP88857194 SP88857190 SP88850190
10790-828EA 3607.77 CAD
10790-828 10791-528 10791-594
Super Nuova+™ Hot Plate Stirrer, Thermo Scientific
Stirrers Magnetic Hotplate Stirrers
Thermo Scientific™ SuperNuova+™ stirring hotplate performs with dependable safety and simple operation, units are available in two sizes including a multi-position model.
  • Top plate choices in ceramic or aluminum
  • Easy-to-read LED display for heating and stirring
  • Raised display design protects electronics from spills
  • Ability to set programs (ramp/dwell) as well as save settings
  • Optional PT1000 temperature probe and splash guard protection shield
Stir trac features for slow-speed stirring, consistent speed control, and stronger magnetic coupling. HotMore Product Information