Insterprep® Fraser Broth, Seward
Supplier: Seward Laboratory System
INS-2102 INS-2202 INS-2302
10025-022PK 1231.77 CAD
10025-022 10818-616 10818-618
Insterprep® Fraser Broth, Seward
Media for Microbiology Prepared Media for Microbiology
Insterprep® is a new range of instantly soluble, sterile, granulated media specifically designed to speed up and simplify sample preparation for microbiological analysis
These instantly soluble, sterile Fraser Broth media granules are supplied ready to use. Choose the Insterprep® sachet to fit specific protocol needs, or the Stomacher® bag. Simply add sterile distilled water and the sample as per protocol.
The Insterprep® pre-weighed, sterilized media bags are packaged with the correct amount of granular media, ready for instant use. The Insterprep® range of products is designed to help forward thinking laboratories cut waste and costs, manage laboratory resources more efficiently, and produce dependable and accurate results.