Insterprep® Buffered Peptone Water, Seward
Supplier: Seward
INS-2300 INS-2100 INS-2200
10025-054PK 863.08 CAD
10025-054 10818-614 10818-612
Insterprep® Buffered Peptone Water, Seward
Media for Microbiology Prepared Media for Microbiology
Insterprep® is a new range of instantly soluble, sterile, granulated media specifically designed to speed up and simplify sample preparation for microbiological analysis
These instantly soluble, sterile Buffered Peptone Water media granules are supplied ready to use. Choose the Insterprep® sachet to fit specific protocol needs, or the Stomacher® bag. Simply add sterile distilled water and the sample as per protocol.
The Insterprep® pre-weighed, sterilized media bags are packaged with the correct amount of granular media, ready for instant use. The Insterprep® range of products is designed to help forward thinking laboratories cut waste and costs, manage laboratory resources more efficiently, and produce dependable and accurate results.