TC Crile-Wood Left Handed Needle Holder, OR Grade, Sklar
Supplier: Sklar
21-9030 21-9035
10195-196EA 629.77 CAD
10195-196 10195-198
TC Crile-Wood Left Handed Needle Holder, OR Grade, Sklar
Forceps
Sklar's TC Crile-Wood Left-Handed Needle Holder is a instrument specially designed for left-handed users to hold and guide small to medium size needles and suture materials
The Crile-Wood Needle Holder has a gently tapered, blunt tip and is generally more delicate than the Mayo-Hegar but slightly less delicate than Webster, Halsey, or DeBakey needle holders. The ratcheted finger ring handle aids in securing needles for both continuous and interrupted closures. Tungsten Carbide instruments are designed to stay sharp for a significantly longer period of time than standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.