Hydrion pH Buffers, Micro Essential Lab
Supplier: MICRO ESSENTIAL LABORATORY I
2801000
2801100
2801200
280200
280400
280500
280600
280640
280686
280700
2807400
280800
280900
CC-725
280-3
34175-077PK
30.29
CAD
34175-077
34175-079
34175-081
34175-083
34175-087
34175-089
34175-091
34175-093
34175-095
34175-097
34175-099
60775-984
60775-986
60785-007
62000-406
Hydrion pH Buffers, Micro Essential Lab
Buffers
pH Calibration Buffers
Chemvelopes are made of a triple protective foil consisting of aluminum, polyethylene and paper which keeps the Hydrion Buffers dry and free-flowing.
Hydrion buffer chemvelopes produce 500 ml of pH buffer standard solution when dissolved in distilled water.
Packaging: Each carton contains five chemvelopes.