AIRLESS-WARE® Straus Flasks, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
213210-1000 213210-0500
KT213210-1000EA 501.63 CAD
KT213210-1000 KT213210-0500
AIRLESS-WARE® Straus Flasks, Kimble Chase
Flasks Round Bottom Flasks
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
  • Dry, deoxygenated and grease-free solvents are maintained in this state for relatively long periods of time
  • Quantities of solvent are withdrawn from the flask using either a cannula or a syringe with a proper length needle
  • Valve is 0-4 mm and has an plug
Solvent integrity is provided by the KONTES® HI-VAC® valve, which provides access to the flask contents.