Disposable Serological Pipettes, Glass, Sterile, TD Color-Coded, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
72102-11100 72102-21100 72102-5110 72102-10110
89085-762CS 626.2 CAD
89085-762 89085-764 89085-766 89085-768
Disposable Serological Pipettes, Glass, Sterile, TD Color-Coded, Kimble Chase
Pipettes Serological Pipettes
Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements.
  • Non-pyrogenic
  • Pipettes have a uniform pulled tip and are clean, plugged, sterile and ready for use
  • Calibrated To Deliver (TD) to the tip
  • Provided with 'negative' graduations to increase the versatility of the pipet
  • Designed from ASTM Specification E714
Ordering information: Packed in a convenient recloseable canister pack containing 50 pieces sealed in a plastic bag.