DuPont™ ProShield® 6 SFR Coveralls with Standard Hood
Supplier: DuPont
TM127SBUMD002500 TM127SBUXL002500 TM127SBU2X002500 TM127SBU3X002500 TM127SBU4X002500 TM127SBU5X002500 TM127SBU6X002500 TM127SBU7X002500 TM127SBULG002500
CA13452-972CS 1489.1 CAD
CA13452-972 CA13452-976 CA13452-978 CA13452-980 CA13452-982 10016-566 10016-568 10016-570 10016-572
DuPont™ ProShield® 6 SFR Coveralls with Standard Hood
Disposable Garments
ProShield® 6 SFR is a cost-efficient disposable overgarment designed to help protect and preserve primary flame resistant garments.
  • Standard fit hood
  • Elastic wrists and ankles
  • Front zipper closure
  • Serged seams
ProShield® 6 SFR garments are intended to be worn over primary flame resistant garments to provide a barrier against non-hazardous particles and aerosols. ProShield® 6 SFR garments are designed to not contribute to additional burn injury in the event of an industrial fire or electric arc event. ProShield® 6 SFR garments will not ignite andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Gender
Garment application Industrial,Chemical protection,Laboratory Industrial,Chemical protection,Laboratory Industrial,Chemical protection,Laboratory